Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Brand:  Thermo Scientific™ SH31188.01
 View more versions of this product
Product Code. 17958221
  • 5010.00 € / Pack of 5
Explore more special offers
Description
Description
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 L
Bioprocess Container (BPC)
Horizontally-gusseted