Learn More
Thermo Scientific™ HyperREZ™ XP Carbohydrate Pb2+ HPLC Columns
HyperREZ XP Carbohydrate Pb2+ LC Columns are used for the appropriate counter-ion to meet application requirements.
Brand:  Thermo Scientific™ 69108-057726
 View more versions of this product
Product Code. 11527531
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Manufactured with a proprietary process and subjected to stringent quality control procedures,Thermo Scientific™ HyperREZ™ XP Carbohydrate Pb2+ LC Columns are used for the appropriate counter-ion to meet application requirements. Each column is individually tested to ensure column-to-column reproducibility.
Polymer-based columns for carbohydrate analysis offer benefits to the chromatographer including temperature and pH stability, as well as enhanced solvent choices.
  • Designed for the determination of carbohydrates, saccharides, organic acids and alcohols
  • Efficient and reproducible monodisperse particles
  • Stable for long column lifetimes even at low pH and high temperatures
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Guard Column
LC/MS
725 psi (50 bar)
1 to 11
L34
Spherical, Microporous Polymer
Ligand Exchange
7.7 mm
50 mm
8 μm
90°C
Pb2+ Counter Ion
Carbohydrate Analysis
Guard Column