Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Vacuum Gauge
Order the vacuum gauge if a replacement is needed to measure vacuum in the glass block manifold.
Brand:  Thermo Scientific™ 60104-261
370.53 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10301245
  • 372.38 € / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Order the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Vacuum Gauge if you need a replacement. The manifold enables processing of 16 or 24 samples through HyperSep SPE solid phase extraction cartridges under vacuum. The gauge is available separately or with the valve assembly.
HyperSep Glass Block Manifolds are durable, chemically resistant units designed to provide years of trouble-free extractions.
Polypropylene racks are highly resistant to chemical degradation and abuse, and are supplied according to the number of positions on the manifold lid
Vacuum control valve allows optimization of sample flow rates during extraction
Test tube racks allow the use of 12-13mm or 16mm disposable test tubes
Vacuum gauge and bleed valve enables easy monitoring of the vacuum process
Corner leg supports prevent damage to the luers and tips when the lid is not being used
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuum Gauge & valve assembly, 1 pk
Vacuum Manifold
Vacuum Gauge and Valve Assembly
SPE Sample Preparation
HyperSep
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Vacuum Gauge > Solid phase extraction; For use with hyperSep manifolds