Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Teflon Tip
Use these tips to direct the flow of extracts from the SPE cartridge to the collection tubes of the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold.
Brand: Thermo Scientific™ 60104-245
Description
Use the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Tips to direct the flow of extract from the HyperSep solid phase extraction SPE cartridges to the collection tubes. They are available in packs of 12.
Specifications
Teflon Tips for Glass Block Vacuum Manifold, 12 pk | |
Glass Block Vacuum Manifold | |
Teflon Tips |
SPE | |
HyperSep |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.