missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersep™ Hypercarb™ Fixed Well Plate

Unique material for retention of highly polar compounds.

550.32 € - 704.71 €

Specifications

For Use With (Equipment) Universal Vacuum Manifold
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HypeSep Hypercarb
Type 96-well Plate
View More Specs

Products 2
Product Code Brand Bed Weight Price Quantity & Availability  
Product Code Brand Bed Weight Price Quantity & Availability  
13496976
View Documents
Thermo Scientific™
60306-502
25 mg
550.32 €
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
 
13406986
View Documents
Thermo Scientific™
60306-503
50 mg
704.71 €
Each
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ HyperSep™ and Hypercarb™ SPE contains flat,100% porous graphitic carbon (PGC) with selectivity for structurally similar compounds, offering separation of compounds with simple solvents

  • pH stable 0 to 14
  • High batch-to-batch reproducibility
  • Strong retention properties allow use of low bed weights for concentrated extracts
  • Interaction mechanism with polar molecules
Specifications

Specifications

Universal Vacuum Manifold
HyperSep
96-well Plate
40 to 60 μm
HypeSep Hypercarb
1/Ea
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.