missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ HyperSep™ Polystyrene Lab Plates
Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.
Brand: Thermo Scientific™ 60110-209
This item is not returnable.
View return policy
Description
Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:
- Tissue culture and separation of products
- Concentration of small-scale samples
- Sample clean-up
- Collection of sample after chromatography
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
SAX; Polystyrene | |
40 to 60 μm | |
HyperSep SAX |
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules | |
HyperSep | |
96-Well Plate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction