Learn More
Thermo Scientific™ HyperSep™ PFAS Cartridges
HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination.
252.10 € - 368.46 €
Product Code | Brand | Bed Weight | Description | For Use With (Application) | Particle Size | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Description | For Use With (Application) | Particle Size | Price | Quantity & Availability | ||
18415682
|
Thermo Scientific™
60107-150-6-WAX |
150 mg | HyperSep PFAS SPE, 150 mg, 6 mL, WAX | EPA method 1633, extraction of PFAS from water, tissue, soil, or biosolids | 55 to 65 μm |
252.10 €
Pack of 30 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18465672
|
Thermo Scientific™
60107-20050-WAXGCB |
250 mg | 200 mg Polymeric WAX on top of 50 mg of graphitized carbon black (GCB) | EPA method 1633, extraction of PFAS from water | 55 to 65 μm and 120/400 mesh |
344.22 €
Pack of 30 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18485672
|
Thermo Scientific™
60107-50200-GCBWAX |
250 mg | HyperSep PFAS SPE, Dual PHASE, 50 mg GCB, 200 MG WAX, 6 mL | EPA method 1633, extraction of PFAS from tissue, soil, or biosolids | 55 to 65 μm and 120/400 mesh |
344.22 €
Pack of 30 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18475672
|
Thermo Scientific™
60107-500-6-HLD |
500 mg | HyperSep PFAS SPE, 500 mg, 6 mL, HLD | EPA method 537, extraction of PFAS from water | 20 to 40 μm |
252.10 €
Pack of 30 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18435682
|
Thermo Scientific™
60107-500-6-WAX |
500 mg | HyperSep PFAS SPE, 500 mg, 6 mL, WAX | EU and other regulations, extraction of PFAS from water | 55 to 65 μm |
368.46 €
Pack of 30 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18425682
|
Thermo Scientific™
60107-30-3-WAX |
30 mg | HyperSep PFAS SPE, 30 mg, 3 mL, WAX | EU and other regulations, extraction of PFAS from water | 55 to 65 μm |
268.59 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18495672
|
Thermo Scientific™
60107-200-6-WAX |
200 mg | HyperSep PFAS SPE, 200 mg, 6 mL, WAX | EPA method 1633 (alternative), extraction of PFAS from water, tissue, soil, or biosolids | 55 to 65 μm |
256.96 €
Pack of 30 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
18405682
|
Thermo Scientific™
GCB-10 |
10 g | HyperSep LOOSE CARBON | EPA method 1633, extraction of PFAS from water, tissue, soil, or biosolids in conjunction with SPE cartridge | 55 to 65 μm and 120/400 mesh |
252.10 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
HyperSep SPE PFAS SPE products are designed to address the needs of extraction and concentration of PFAS samples for targeted and not targeted screening while minimizing background contamination. The range of selectivity’s and formats are designed to meet the requirements for main environmental testing methods by extracting and concentrating samples from drinking water, ground water, surface water, wastewater, soil, food and other complex matrices.
Per- and polyfluorinated alkyl substances (PFAS) are among the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances is rapidly growing. SPE solutions are a fundamental part of the analytical workflow for analysis of PFAS compounds.
Thermo Scientific provides a range of products to address PFAS testing requirements e.g. EPA methods: 1633, 537.1, 533.
Specifications
HyperSep | |
Solid Phase Extraction Column |