Learn More
Thermo Scientific™ HyperSep™ Retain AX Cartridges
Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX SPE Cartridges.
127.00 € - 681.72 €
Specifications
For Use With (Application) | Retention of Acidic Analytes |
---|---|
For Use With (Equipment) | Thermo Scientific HyperSep-96 System |
Particle Size | 30 to 50 μm |
Product Line | HyperSep |
Sorbent | HyperSep Quaternary Amine-Modified Polystyrene DVB |
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | ||||
10635535
|
Thermo Scientific™
60107-401 |
30 mg | 1 mL |
227.20 €
Pack of 100 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
10772917
|
Thermo Scientific™
60107-402 |
30 mg | 3 mL | N/A | |||||
10635725
|
Thermo Scientific™
60107-403 |
60 mg | 3 mL |
141.56 €
Pack of 50 |
Estimated Shipment: 10-12-2024
Log in to see stock. |
||||
10117524
|
Thermo Scientific™
60107-404 |
200 mg | 3 mL |
288.10 €
Pack of 50 |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
10595625
|
Thermo Scientific™
60107-405 |
500 mg | 3 mL |
512.19 €
Pack of 50 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
10075934
|
Thermo Scientific™
60107-406 |
500 mg | 6 mL |
334.52 €
Pack of 30 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
||||
10279904
|
Thermo Scientific™
60107-407 |
100 mg | 6 mL |
180.22 €
Pack of 30 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
10055934
|
Thermo Scientific™
60107-408 |
60 mg | 6 mL |
171.91 €
Pack of 30 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
10046504
|
Thermo Scientific™
60107-411 |
150 mg | 6 mL |
189.69 €
Pack of 30 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
10056504
|
Thermo Scientific™
60107-412 |
200 mg | 6 mL |
187.23 €
Pack of 30 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
||||
10260245
|
Thermo Scientific™
60107-414 |
2000 mg | 25 mL |
681.72 €
Pack of 20 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
||||
10270245
|
Thermo Scientific™
60107-415 |
1000 mg | 25 mL |
398.45 €
Pack of 20 |
In Stock
Log in to see stock. |
||||
12106590
|
Thermo Scientific™
60107-417 |
100 mg | 15 mL |
186.14 €
Pack of 50 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||||
13466936
|
Thermo Scientific™
60107-419 |
500 mg | 3 mL |
263.20 €
Pack of 20 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
||||
Description
Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with quaternary amine functional groups. Their use in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Widely used in forensic and clinical analysis of THC and its metabolites, Hypersep Retain SPE cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.- High, consistent recoveries
- Ideal for large samples
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- pH stable 0 to 14
- Available in a wide range of chemistries and bed weights
Specifications
Retention of Acidic Analytes | |
30 to 50 μm | |
HyperSep Quaternary Amine-Modified Polystyrene DVB |
Thermo Scientific HyperSep-96 System | |
HyperSep | |
Solid Phase Extraction Cartridge |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.