Learn More
Thermo Scientific™ HyperSep™ SAX Plates
Obtain excellent performance for extraction of carboxylic and other weak acids with these strong anion exchanger SPE well plates and individual wells.
353.00 € - 462.17 €
Specifications
For Use With (Application) | Extraction of Weak Acids |
---|---|
Particle Size | 40 to 60 μm |
Product Line | HyperSep |
Sorbent | HyperSep SAX |
Type | 96-well Plate |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10457894
|
Thermo Scientific™
60300-565 |
SAX 10 mg/1 mL 96 Removable Well Plate, 1 pk | N/A | ||||||
10427073
|
Thermo Scientific™
60300-567 |
SAX 50 mg/1 mL 96 Removable Well Plate, 1 pk |
460.97 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
10085674
|
Thermo Scientific™
60300-568 |
SAX 100 mg/1 mL 96 Removable Well Plate, 1 pk | N/A | ||||||
13407006
|
Thermo Scientific™
60307-242 |
SAX 25 mg/2 mL 96 Fixed Well Plate, 1 pk |
462.17 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
13486996
|
Thermo Scientific™
60307-243 |
SAX 50 mg/2 mL 96 Fixed Well Plate, 1 pk |
460.97 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ SAX Plates feature a strong anion exchange sorbent for extraction of weak acids. Typical applications include:
- Isolation of anionic proteins
- Removal of acidic food pigments
- Isolation of phenolic compounds
- Nucleic acids, nucleotides and surfactants
- Amine group masks the effect of the carbon chain of the functional group
- Selectivity can be tuned by choice of buffer
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Extraction of Weak Acids | |
HyperSep | |
96-well Plate |
40 to 60 μm | |
HyperSep SAX | |
1 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.