Learn More
Thermo Scientific™ HyperSep™ SAX Cartridges
Obtain excellent performance for extraction of carboxylic and other weak acids with these strong anion exchanger SPE cartridges.
183.33 € - 490.91 €
Specifications
For Use With (Application) | Extraction of Weak Acids |
---|---|
For Use With (Equipment) | Thermo Scientific HyperSep-96 System |
Particle Size | 40 to 60 μm |
Product Line | HyperSep |
Sorbent | HyperSep SAX |
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | ||||
10100693
|
Thermo Scientific™
60108-360 |
500 mg | 6 mL |
206.34 €
Pack of 30 |
In Stock
Log in to see stock. |
||||
10575615
|
Thermo Scientific™
60108-417 |
50 mg | 1 mL |
253.83 €
Pack of 100 |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||||
10130743
|
Thermo Scientific™
60108-418 |
100 mg | 1 mL |
259.82 €
Pack of 100 |
Estimated Shipment: 11-02-2025
Log in to see stock. |
||||
10289414
|
Thermo Scientific™
60108-419 |
200 mg | 3 mL |
183.33 €
Pack of 50 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
10045724
|
Thermo Scientific™
60108-434 |
1000 mg | 6 mL |
200.62 €
Pack of 30 |
Estimated Shipment: 14-01-2025
Log in to see stock. |
||||
10786134
|
Thermo Scientific™
60108-521 |
500 mg | 3 mL |
217.91 €
Pack of 50 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||||
10574465
|
Thermo Scientific™
60108-713 |
2000 mg | 15 mL |
205.23 €
Pack of 20 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
10651006
|
Thermo Scientific™
60108-714 |
5000 mg | 25 mL |
352.02 €
Pack of 20 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
10107714
|
Thermo Scientific™
60108-715 |
10 g | 75 mL |
490.91 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ SAX Cartridges feature a strong anion exchange sorbent for extraction of weak acids. Typical applications include:
- Isolation of anionic proteins
- Removal of acidic food pigments
- Isolation of phenolic compounds
- Nucleic acids, nucleotides and surfactants
- Amine group masks the effect of the carbon chain of the functional group
- Selectivity can be tuned by choice of buffer
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Extraction of Weak Acids | |
40 to 60 μm | |
HyperSep SAX |
Thermo Scientific HyperSep-96 System | |
HyperSep | |
Solid Phase Extraction Cartridge |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.