Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Empty Well Plates
These empty 96-well plates are used with Thermo Scientific™ HyperSep 96-well plate applications.
Brand:  Thermo Scientific™ 60300-301
 View more versions of this product
Product Code. 10007734
  • 36.53 € / Pack of 40
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format.
  • Available in a wide range of chemistries and bed weights
  • Ideal for large samples and conform to industry standard configurations
  • Allows for efficient routine processing by vacuum, by positive pressure or centrifugation.
  • The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility.
  • The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation.
  • Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
  • HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 mL
Solid Phase Extraction
96-well SPE Plate