Learn More
Thermo Scientific™ HyperSep™ Verify AX Cartridges
Get improved analysis of acidic drugs and metabolites with these SPE cartridges featuring both nonpolar and ionic separation characteristics.
171.79 € - 347.23 €
Specifications
For Use With (Application) Analysis of Acidic Drugs and Metabolites
For Use With (Equipment) Thermo Scientific HyperSep-96 System
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep Reversed Phase C8/Quaternary Amine Ion Exchanger
View More Specs
Products 8
Product Code Brand Bed Weight Column Capacity (Metric) Price Quantity & Availability  
Product Code Brand Bed Weight Column Capacity (Metric) Price Quantity & Availability  
11547591
View Documents
Thermo Scientific™
60108-727
130 mg 1 mL
347.23 €
Pack of 100
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
10445923
View Documents
Thermo Scientific™
60108-728
300 mg 3 mL
207.14 €
Pack of 50
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
10584495
View Documents
Thermo Scientific™
60108-729
500 mg 3 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10506205
View Documents
Thermo Scientific™
60108-730
200 mg 6 mL
213.30 €
Pack of 50
Estimated Shipment: 23-12-2024
to see stock.
Add to basket
10203083
View Documents
Thermo Scientific™
60108-731
500 mg 6 mL
215.51 €
Pack of 50
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
10639285
View Documents
Thermo Scientific™
60108-732
1000 mg 6 mL
213.13 €
Pack of 30
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
11587671
View Documents
Thermo Scientific™
60108-764
200 mg 10 mL
171.79 €
Pack of 50
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
13486936
View Documents
Thermo Scientific™
60108-767
130 mg 10 mL
229.89 €
Pack of 50
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
Description
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ Verify AX Cartridges feature both nonpolar and ionic separation chemistries for improved sample preparation in the analysis of acidic drugs and metabolites from biological matrices. They are especially well-suited for screening and analysis of THC and its metabolites.
The sorbent used in HyperSep Verify AX Cartridges is based on two functional groups: reversed phase C8 and a quaternary amine ion exchanger. The chemistry is co-polymerized on a rigid, purified silica gel support.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Specifications
Analysis of Acidic Drugs and Metabolites
40 to 60 μm
HyperSep Reversed Phase C8/Quaternary Amine Ion Exchanger
Thermo Scientific HyperSep-96 System
HyperSep
Solid Phase Extraction Cartridge
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.