missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ APS-2 Amino HPLC Column

Analyze sugars with this versatile amino propyl phase of Thermo Scientific™ Hypersil™ APS-2 Amino LC Columns.

Brand:  Thermo Scientific™ 30710-254630

 View more versions of this product

Product Code. 11537491

  • 667.81 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Extra sensitivity for sugars
  • Can be used in three modes: reversed phase, normal phase, weak anion exchange
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4.6 mm
10 μm
120 Å
L8
Silica
Mixed Mode
250 mm
2.5 to 8
170 m²/g
Amino (NH2)
2%
Reversed Phase, Normal Phase, Ion Exchange, HILIC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers