Learn More
Thermo Scientific™ Hypersil GOLD™ Phenyl HPLC Columns
Use for quick analysis of aromatic analytes, including glucocorticosteroids, estrogens, fat-soluble vitamins, and phospholipids.
346.00 € - 2717.06 €
Specifications
pH 2 to 8
Pore Size 175 Å
Surface Area 220 m²/g
Temperature 60°C
USP Type L11
View More Specs
Products 27
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Column Format Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Column Format Price Quantity & Availability  
16398576
View Documents
Thermo Scientific™
25905-259070A
250 mm 10 mm 5 μm Preparative Column, Standard
2717.06 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
10675335
View Documents
Thermo Scientific™
25905-052130
50 mm 2.1 mm 5 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10762907
View Documents
Thermo Scientific™
25905-054630
50 mm 4.6 mm 5 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10753477
View Documents
Thermo Scientific™
25905-102130
100 mm 2.1 mm 5 μm Analytical Column
764.03 €
Each
In Stock
to see stock.
Add to basket
10127604
View Documents
Thermo Scientific™
25905-103030
100 mm 3 mm 5 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10753667
missing translation for 'viewDocuments'
Thermo Scientific™
25905-104630
100 mm 4.6 mm 5 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10014574
missing translation for 'viewDocuments'
Thermo Scientific™
25905-154630
150 mm 4.6 mm 5 μm Analytical Column
766.18 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Zum Warenkorb hinzufügen
10005784
missing translation for 'viewDocuments'
Thermo Scientific™
25905-254030
250 mm 4 mm 5 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10442595
missing translation for 'viewDocuments'
Thermo Scientific™
25905-254630
250 mm 4.6 mm 5 μm Analytical Column
807.26 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10351445
missing translation for 'viewDocuments'
Thermo Scientific™
25905-014001
10 mm 4 mm 5 μm Guard Cartridge, 4 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10525435
missing translation for 'viewDocuments'
Thermo Scientific™
25905-013001
10 mm 3 mm 5 μm Guard Cartridge, 4 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10025644
missing translation for 'viewDocuments'
Thermo Scientific™
25905-012101
10 mm 2.1 mm 5 μm Guard Cartridge, 4 Pk.
431.44 €
Pack of 4
Estimated Shipment: 22-01-2025
to see stock.
Zum Warenkorb hinzufügen
10287924
missing translation for 'viewDocuments'
Thermo Scientific™
25903-154630
150 mm 4.6 mm 3 μm Analytical Column
840.75 €
Each
Estimated Shipment: 15-01-2025
to see stock.
Zum Warenkorb hinzufügen
10446984
missing translation for 'viewDocuments'
Thermo Scientific™
25903-153030
150 mm 3 mm 3 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10277924
missing translation for 'viewDocuments'
Thermo Scientific™
25903-152130
150 mm 2.1 mm 3 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10794627
missing translation for 'viewDocuments'
Thermo Scientific™
25903-151030
150 mm 1 mm 3 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10690256
missing translation for 'viewDocuments'
Thermo Scientific™
25903-104630
100 mm 4.6 mm 3 μm Analytical Column
799.69 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Zum Warenkorb hinzufügen
10437324
missing translation for 'viewDocuments'
Thermo Scientific™
25903-103030
100 mm 3 mm 3 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10056364
missing translation for 'viewDocuments'
Thermo Scientific™
25903-102130
100 mm 2.1 mm 3 μm Analytical Column
836.43 €
Each
Estimated Shipment: 15-01-2025
to see stock.
Zum Warenkorb hinzufügen
10208264
missing translation for 'viewDocuments'
Thermo Scientific™
25903-052130
50 mm 2.1 mm 3 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10026124
missing translation for 'viewDocuments'
Thermo Scientific™
25903-014001
10 mm 4 mm 3 μm Guard Cartridge, 4 Pk.
452.82 €
Pack of 4
In Stock
to see stock.
Zum Warenkorb hinzufügen
10279654
missing translation for 'viewDocuments'
Thermo Scientific™
25903-013001
10 mm 3 mm 3 μm Guard Cartridge, 4 Pk.
473.87 €
Pack of 4
Estimated Shipment: 16-01-2025
to see stock.
Zum Warenkorb hinzufügen
10380915
missing translation for 'viewDocuments'
Thermo Scientific™
25903-012101
10 mm 2.1 mm 3 μm Guard Cartridge, 4 Pk.
444.15 €
Pack of 4
Estimated Shipment: 15-01-2025
to see stock.
Zum Warenkorb hinzufügen
10595125
missing translation for 'viewDocuments'
Thermo Scientific™
25902-202130
200 mm 2.1 mm 1.9 μm Analytical Column This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10431825
missing translation for 'viewDocuments'
Thermo Scientific™
25902-152130
150 mm 2.1 mm 1.9 μm Analytical Column
1101.61 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10207834
missing translation for 'viewDocuments'
Thermo Scientific™
25902-102130
100 mm 2.1 mm 1.9 μm Analytical Column
992.60 €
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10422015
missing translation for 'viewDocuments'
Thermo Scientific™
25902-052130
50 mm 2.1 mm 1.9 μm Analytical Column
1002.92 €
Each
Estimated Shipment: 15-01-2025
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Get excellent retention and unique selectivity for aromatic and moderately polar analytes with Thermo Scientific™ Hypersil GOLD™ Phenyl reversed phase HPLC columns. A butyl (C4) linker in the phenyl bonded phase allows for superior alignment of the phenyl ring with aromatic molecules, thus enhancing pi-pi interactions and improving the retention of the molecules. The C4 linker also provides the stationary phase with moderate hydrophobicity, making it ideal for the separation of analyte mixtures with varying polarity and aromaticity. The outstanding peak shape results in greater sensitivity.
Special Selectivity
  • Provides selectivity complementary to that provided by alkyl chain phases
  • Moderate hydrophobicity compared to C18
About the Hypersil GOLD Family of HPLC Columns
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3, and 5 μm particle sizes for all your separation needs
Spezifikation
Spezifikation
2 to 8
220 m²/g
L11
Yes
175 Å
60°C
Reversed Phase
Reversed Phase
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt