Learn More
Thermo Scientific™ Hypersil GOLD™ PREP aQ HPLC Columns
Stable in 100% aqueous mobile phase, Hypersil GOLD aQ polar endcapped C18 preparative HPLC columns provide enhanced retention and resolution of polar analytes.
251.14 € - 32074.39 €
Specifications
Particle Size | 5 μm |
---|---|
pH | 2 to 9 |
Pore Size | 175 Å |
Surface Area | 220 m²/g |
Temperature | 60°C |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Column Format | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Column Format | Price | Quantity & Availability | |||
16378546
|
Thermo Scientific™
25305-018823A |
8 mm | 10 mm | Guard Column |
251.14 €
Each |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||
16388546
|
Thermo Scientific™
25305-019023A |
20 mm | 10 mm | Guard Column |
397.88 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
16398546
|
Thermo Scientific™
25305-019323A |
30 mm | 10 mm | Guard Column, 1/16 in. |
550.27 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16308556
|
Thermo Scientific™
25305-019323B |
30 mm | 10 mm | Guard Column, 1/8 in. |
550.27 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16318556
|
Thermo Scientific™
25305-029023A |
20 mm | 20 mm | Guard Column | N/A | ||||
16328556
|
Thermo Scientific™
25305-029323A |
30 mm | 20 mm | Guard Column, 1/16 in. |
838.09 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16338556
|
Thermo Scientific™
25305-029323B |
30 mm | 20 mm | Guard Column, 1/8 in. |
838.09 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16348556
|
Thermo Scientific™
25305-109070A |
100 mm | 10 mm | Preparative Column, Standard |
1960.01 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||
16358556
|
Thermo Scientific™
25305-109270A |
100 mm | 10 mm | Preparative Column, Standard |
3807.40 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16368556
|
Thermo Scientific™
25305-109270XL |
100 mm | 25 mm | Preparative Column, Xtended Life |
7158.43 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16378556
|
Thermo Scientific™
25305-109370A |
100 mm | 30 mm | Preparative Column, Standard |
7446.65 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16388556
|
Thermo Scientific™
|
100 mm | 30 mm | Preparative Column, Xtended Life | N/A | ||||
16398556
|
Thermo Scientific™
25305-109570A |
100 mm | 50 mm | Preparative Column, Standard |
13499.20 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16308566
|
Thermo Scientific™
25305-109570XL |
100 mm | 50 mm | Preparative Column, Xtended Life |
19827.58 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16318566
|
Thermo Scientific™
25305-159070A |
150 mm | 10 mm | Preparative Column, Standard |
2238.85 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16328566
|
Thermo Scientific™
25305-159270A |
150 mm | 20 mm | Preparative Column, Standard |
4271.60 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16338566
|
Thermo Scientific™
25305-159270XL |
150 mm | 25 mm | Preparative Column, Xtended Life |
8102.38 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16348566
|
Thermo Scientific™
25305-159370A |
150 mm | 30 mm | Preparative Column, Standard |
9720.52 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||
16358566
|
Thermo Scientific™
|
150 mm | 30 mm | Preparative Column, Xtended Life | N/A | ||||
16368566
|
Thermo Scientific™
25305-159570A |
150 mm | 50 mm | Preparative Column, Standard |
18804.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16378566
|
Thermo Scientific™
|
150 mm | 50 mm | Preparative Column, Xtended Life | N/A | ||||
16388566
|
Thermo Scientific™
25305-259070A |
250 mm | 10 mm | Preparative Column, Standard |
2630.85 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||
16398566
|
Thermo Scientific™
25305-259270A |
250 mm | 20 mm | Preparative Column, Standard |
5253.62 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16308576
|
Thermo Scientific™
25305-259270XL |
250 mm | 25 mm | Preparative Column, Xtended Life |
10027.85 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||
16318576
|
Thermo Scientific™
25305-259370A |
250 mm | 30 mm | Preparative Column, Standard |
12258.43 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16328576
|
Thermo Scientific™
|
250 mm | 30 mm | Preparative Column, Xtended Life | N/A | ||||
16338576
|
Thermo Scientific™
25305-259570A |
250 mm | 50 mm | Preparative Column, Standard |
32074.39 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||
16348576
|
Thermo Scientific™
|
250 mm | 50 mm | Preparative Column, Xtended Life | N/A | ||||
Description
Retain and resolve polar analytes for your scale up purification needs by using Thermo Scientific™ Hypersil GOLD™ aQ Polar Endcapped C18 preparative columns. These C18 preparative HPLC columns are perfect for challenging reversed phase separations using highly aqueous mobile phases. Our columns come in a wide range of dimensions and an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Alternative selectivity with polar endcapped C18 stationary phase
- Polar functional group in endcap provides further interaction mechanism for separation of polar compounds.
- Yields superior retention of polar compounds.
- C18 (USP L1) selectivity.
Stability in highly aqueous mobile phases
- Applicable in 100% aqueous mobile phases.
- No loss of performance or stability.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
- Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
- Standard threaded hardware for column i.d. < 40mm.
- Standard flange hardware for column i.d. ≥ 40mm.
- Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
- Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
- Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
- If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
- Please contact your local sales representative for Customer Specific Orders.
- Custom built columns are not eligible for returns.
About the Hypersil GOLD family of HPLC columns
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
Specifications
5 μm | |
175 Å | |
60°C | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
2 to 9 | |
220 m²/g | |
L1 | |
12% | |
Polar |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.