missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil GOLD™ AX HPLC Columns

Product Code. 10469644 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
10 mm
30 mm
50 mm
100 mm
150 mm
200 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
4 mm
4.6 mm
Particle Size:
1.9 μm
3 μm
5 μm
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 10469644

Brand: Thermo Scientific™ 26105012101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns.

Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns. The weak anion-exchange phase, created by a polymeric amine ligand bonded to highly pure base-deactivated silica, separates multiple charged analytes. The bonded phase offers additional chromatographic options, including hydrophilic interaction chromatography (HILIC), which is ideal for the separation of sugars and other highly polar species.

Excellent Chromatographic Performance
  • Outstanding peak shape and selectivity
  • Superb stability under demanding pH conditions
  • Higher efficiency than polymer-based ion exchange columns
  • Improved speed with 1.9μm particle size columns
About the Hypersil GOLD Family of HPLC Columns



Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5μm particle sizes for all your separation needs
TRUSTED_SUSTAINABILITY

Specifications

Column Format Guard Cartridge, 4 Pk.
Diameter (Metric) 2.1 mm
Flow Rate 0.2 mL/min
For Use With Uniguard Direct-Connection and Stand Alone Holder
Length (Metric) 10 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 175 Å
Surface Area 220 m²/g
Temperature 60°C
Stationary Phase Polymeric Amine
Product Line Hypersil GOLD
Packing Material Base-Deactivated Silica
Carbon Load 6%
Column Type Mixed Mode
Endcapped No
Phase HILIC and Ion Exchange
Type Guard Cartridge, 4 Pk.
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.