Learn More
Thermo Scientific™ Hypersil™ PREP ODS C18 HPLC Columns
Perfect for routine workflows, our Hypersil PREP ODS preparative LC column is a general purpose C18 used in a broad range of USP L1 applications.
149.54 € - 17229.92 €
Specifications
pH | 2 to 8 |
---|---|
Pore Size | 130 Å |
Surface Area | 195 m²/g |
Temperature | 60°C |
USP Type | L1 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||
16388976
|
Thermo Scientific™
40203-052130 |
50 mm | 2 mm | 3 μm | Preparative Column, Standard |
625.27 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16398976
|
Thermo Scientific™
40203-054630 |
50 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
552.65 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16308986
|
Thermo Scientific™
40203-102130 |
100 mm | 2 mm | 3 μm | Preparative Column, Standard |
681.87 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16318986
|
Thermo Scientific™
40203-104630 |
100 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
678.19 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||
16328986
|
Thermo Scientific™
40203-152130 |
150 mm | 2 mm | 3 μm | Preparative Column, Standard |
759.42 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16338986
|
Thermo Scientific™
40203-154630 |
150 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
761.89 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16348986
|
Thermo Scientific™
40203-252130 |
250 mm | 2 mm | 3 μm | Preparative Column, Standard |
807.43 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16358986
|
Thermo Scientific™
40203-254630 |
250 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
772.96 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||
16368986
|
Thermo Scientific™
40205-052130 |
50 mm | 2 mm | 5 μm | Preparative Column, Standard |
537.88 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16378986
|
Thermo Scientific™
40205-054630 |
50 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
566.19 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||
16388986
|
Thermo Scientific™
40205-059070A |
50 mm | 10 mm | 5 μm | Preparative Column, Standard |
1152.05 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16398986
|
Thermo Scientific™
40205-059270A |
50 mm | 20 mm | 5 μm | Preparative Column, Standard |
1344.07 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16308996
|
Thermo Scientific™
40205-059270XL |
50 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
4361.72 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16318996
|
Thermo Scientific™
40205-102130 |
100 mm | 2 mm | 5 μm | Preparative Column, Standard |
695.42 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16328996
|
Thermo Scientific™
40205-104630 |
100 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
609.27 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16338996
|
Thermo Scientific™
40205-109070A |
100 mm | 10 mm | 5 μm | Preparative Column, Standard |
1191.44 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16348996
|
Thermo Scientific™
40205-109270A |
100 mm | 20 mm | 5 μm | Preparative Column, Standard |
2971.23 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16358996
|
Thermo Scientific™
40205-109270XL |
100 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
6423.44 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16368996
|
Thermo Scientific™
40205-152130 |
150 mm | 2 mm | 5 μm | Preparative Column, Standard |
657.27 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16378996
|
Thermo Scientific™
40205-154630 |
150 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
678.19 €
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||
16388996
|
Thermo Scientific™
40205-159070A |
150 mm | 10 mm | 5 μm | Preparative Column, Standard |
1384.69 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16398996
|
Thermo Scientific™
40205-159270A |
150 mm | 20 mm | 5 μm | Preparative Column, Standard |
3379.86 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||
16309006
|
Thermo Scientific™
40205-159270XL |
150 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
7180.83 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16319006
|
Thermo Scientific™
40205-159370A |
150 mm | 30 mm | 5 μm | Preparative Column, Standard |
5516.59 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16329006
|
Thermo Scientific™
40205-159370XL |
150 mm | 30 mm | 5 μm | Preparative Column, Xtended Life |
9092.24 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16339006
|
Thermo Scientific™
40205-159570A |
150 mm | 50 mm | 5 μm | Preparative Column, Standard |
12116.29 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16349006
|
Thermo Scientific™
40205-159570XL |
150 mm | 50 mm | 5 μm | Preparative Column, Xtended Life |
15986.67 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16359006
|
Thermo Scientific™
40205-252130 |
250 mm | 2 mm | 5 μm | Preparative Column, Standard |
702.81 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16369006
|
Thermo Scientific™
40205-254630 |
250 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
686.80 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
16379006
|
Thermo Scientific™
40205-259070A |
250 mm | 10 mm | 5 μm | Preparative Column, Standard |
1570.18 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||
16389006
|
Thermo Scientific™
40205-259270A |
250 mm | 20 mm | 5 μm | Preparative Column, Standard |
3800.80 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
16399006
|
Thermo Scientific™
40205-259270XL |
250 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
7070.50 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||
16309016
|
Thermo Scientific™
40205-259370A |
250 mm | 30 mm | 5 μm | Preparative Column, Standard |
6591.67 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16319016
|
Thermo Scientific™
40205-259370XL |
250 mm | 30 mm | 5 μm | Preparative Column, Xtended Life |
9971.63 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16329016
|
Thermo Scientific™
40205-259570A |
250 mm | 50 mm | 5 μm | Preparative Column, Standard |
13361.89 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16339016
|
Thermo Scientific™
40205-259570XL |
250 mm | 50 mm | 5 μm | Preparative Column, Xtended Life |
17229.92 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16349016
|
Thermo Scientific™
40210-018823A |
8 mm | 10 mm | 10 μm | Guard Column |
149.54 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||
16359016
|
Thermo Scientific™
40210-019023A |
20 mm | 10 mm | 10 μm | Guard Column |
261.70 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
||
16369016
|
Thermo Scientific™
40210-019323A |
30 mm | 10 mm | 10 μm | Guard 1/16 in. |
317.13 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16379016
|
Thermo Scientific™
40210-019323B |
30 mm | 10 mm | 10 μm | Guard 1/8 in. |
317.13 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16389016
|
Thermo Scientific™
40210-029023A |
20 mm | 20 mm | 10 μm | Guard Column | N/A | |||
16399016
|
Thermo Scientific™
40210-029323A |
30 mm | 20 mm | 10 μm | Guard 1/16 in. |
523.40 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16309026
|
Thermo Scientific™
40210-029323B |
30 mm | 20 mm | 10 μm | Guard 1/8 in. |
523.40 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16319026
|
Thermo Scientific™
40210-059070A |
50 mm | 10 mm | 10 μm | Preparative Column, Standard |
1036.37 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16329026
|
Thermo Scientific™
40210-059270A |
50 mm | 20 mm | 10 μm | Preparative Column, Standard |
1208.68 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16339026
|
Thermo Scientific™
40210-059270XL |
50 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
4211.08 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16349026
|
Thermo Scientific™
40210-109070A |
100 mm | 10 mm | 10 μm | Preparative Column, Standard |
1072.05 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16359026
|
Thermo Scientific™
40210-109270A |
100 mm | 20 mm | 10 μm | Preparative Column, Standard |
2673.36 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16369026
|
Thermo Scientific™
40210-109270XL |
100 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
5472.67 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16379026
|
Thermo Scientific™
40210-159070A |
150 mm | 10 mm | 10 μm | Preparative Column, Standard |
1244.38 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16389026
|
Thermo Scientific™
40210-159270A |
150 mm | 20 mm | 10 μm | Preparative Column, Standard |
3042.62 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16399026
|
Thermo Scientific™
40210-159270XL |
150 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
5855.02 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16309036
|
Thermo Scientific™
40210-159370A |
150 mm | 30 mm | 10 μm | Preparative Column, Standard |
4963.95 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16319036
|
Thermo Scientific™
40210-159370XL |
150 mm | 30 mm | 10 μm | Preparative Column, Xtended Life |
8521.26 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16329036
|
Thermo Scientific™
40210-159570A |
150 mm | 50 mm | 10 μm | Preparative Column, Standard |
10906.38 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16339036
|
Thermo Scientific™
40210-159570XL |
150 mm | 50 mm | 10 μm | Preparative Column, Xtended Life |
15349.99 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16349036
|
Thermo Scientific™
40210-259070A |
250 mm | 10 mm | 10 μm | Preparative Column, Standard |
1412.14 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16359036
|
Thermo Scientific™
40210-259270A |
250 mm | 20 mm | 10 μm | Preparative Column, Standard |
3422.94 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||
16369036
|
Thermo Scientific™
40210-259270XL |
250 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
6248.84 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16379036
|
Thermo Scientific™
40210-259370A |
250 mm | 30 mm | 10 μm | Preparative Column, Standard |
5934.03 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16389036
|
Thermo Scientific™
40210-259370XL |
250 mm | 30 mm | 10 μm | Preparative Column, Xtended Life |
9314.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16399036
|
Thermo Scientific™
40210-259570A |
250 mm | 50 mm | 10 μm | Preparative Column, Standard |
12026.44 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
16309046
|
Thermo Scientific™
40210-259570XL |
250 mm | 50 mm | 10 μm | Preparative Column, Xtended Life |
16508.52 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
Description
Scale up your analytical method using Thermo Scientific™ Hypersil™ PREP ODS C18 reversed phase preparative HPLC Columns. Our columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. This C18 column is a general purpose column used in a broad range of applications, perfect for routine workflows.
For Routine Applications
- C18 (USP L1) selectivity.
- pH stable from 2 to 8.
- Maximum temperature of 60°C.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
- Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
- Standard threaded hardware for column i.d. < 40mm.
- Standard flange hardware for column i.d. ≥ 40mm.
- Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
- Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
- Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
- If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
- Please contact your local sales representative for Customer Specific Orders.
- Custom built columns are not eligible for returns.
Specifications
2 to 8 | |
195 m²/g | |
L1 | |
Spherical, Fully Porous Silica | |
Reversed Phase | |
Reversed Phase |
130 Å | |
60°C | |
C18 | |
10% | |
Yes |