missing translation for 'onlineSavingsMsg'
Learn More
Hypoxanthine, 99.5%, Thermo Scientific Chemicals Product Code.: 10787763

Hypoxanthine, 99.5%, Thermo Scientific Chemicals

Product Code. 10787763
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10787763

Brand: Thermo Scientific Acros 122010050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-94-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00005725
InChI Key FDGQSTZJBFJUBT-UHFFFAOYSA-N
Synonym hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
PubChem CID 790
ChEBI CHEBI:17368
SMILES O=C1N=CNC2=C1NC=N2

Specifications

Melting Point >250.0°C
Color Brown to Colorless
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 26, 416
Merck Index 15, 4908
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in diluted alkalies and acids, practically insoluble in most common organic, solvents
IUPAC Name 3,7-dihydropurin-6-one
Transmittance 95% min. (c=0.1, 0.2M NaOH, 430nm) 1cm cell
Formula Weight 136.11
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Hypoxanthine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451740 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number122010050,122010250,122011000
Lot NumberA0399296 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number122010050,122010250,122011000
Lot NumberA0412228 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number122010050,122010250,122011000
Lot NumberA0447433 Certificate TypeCertificate of Analysis Date24/11/2022 Catalog Number122010050,122010250,122011000
Lot NumberA0439610 Certificate TypeCertificate of Analysis Date04/03/2022 Catalog Number122010050,122010250,122011000
compliance-icons
Product Identifier
  • Hypoxanthine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hypoxanthine, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.