Learn More
STEMCELL Technologies Hypoxia Incubator Chamber
Chamber for generation of a hypoxic environment for tissue culture
Brand:  STEMCELL Technologies 27310
Encompass_Preferred
Product Code. 16226860
  • 1718.55 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The Hypoxia Incubator Chamber is a self-contained and sealed chamber that fits inside existing laboratory incubators
  • Each unit utilizes a surface-type seal in which all portions of the O-ring are uniformly compressed by a ring clamp for a reliable, air-tight seal
  • The cylindrical walls and semi-spherical top and bottom minimize gas flow resistance and eliminate dead space during initial purging
  • The chambers have an integrated stacking feature for storage during or after experimentation
  • All units are molded from high quality polycarbonate and will not break, crack or cloud with extended use
  • Capacity: 84 x 35 mm plates, 27 x 60 mm plates, 12 x 100 mm plates, 12 x 96-well plates, 18 x 25 cm2 flasks
  • Hypoxia Incubator Chamber (Catalog #27310) Components: Lid* (Polycarbonate)
  • Base* (Polycarbonate) with O-ring and Tubing Clamp
  • Tray* (Polycarbonate)
  • Ring Clamp
  • O-ring
  • Tubing Clamp with Tubing
Specifications
Specifications
Cell Culture
1 Chamber