Learn More
Thermo Scientific™ HyPURITY™ C8 HPLC Columns
Use as a general-purpose C8 phase for polar and neutral analytes.
Brand:  Thermo Scientific™ 22205-254030
 View more versions of this product
Product Code. 10149474
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
High quality, highly pure silica columns.
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 mm
5 μm
190 Å
L7
Silica
Reversed Phase
Reversed Phase
250 mm
2 to 9
200 m²/g
C8
8%
Yes