Learn More
Ambion™ mirVana™ miRNA Inhibitor
mirVana™ miRNA Inhibitor
Brand:  Ambion™ 4464092
 View more versions of this product
Product Code. 13403089
  • 27160.00 € / Each
Explore more special offers
Description
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Room temperature
mirVana™ miRNA Inhibitor, 0.25nmol, Small, Chemically Modified Single-Stranded RNA Molecules, Designed to Specifically Bind to and Inhibit Endogenous miRNA, Molecules and Enable miRNA Functional Analysis by Down-Regulation of miRNA Activity
mirVana™ miRNA Inhibitor
0.25 nmol