missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ mirVana™ miRNA Mimic

Small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity.

Brand:  Ambion™ 4464066

319.68 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 13426696

  • 432.00 € / 5nmol

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. mirVana™ miRNA Mimics are more specific than their predecessors due to proprietary chemical modifications.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
miRNA
1 Each
mirVana™ Mimics and Inhibitors, 5nmol, Mimics, Which are More Specific than their Predecessors, miRNA Inhibitors, Which are More Potent than Their Predecessors, Enable miRNA Functional Analysis by Up-regulation of miRNA Activity
mirVana™ Mimics and Inhibitors
5 nmol
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ambion™ mirVana™ miRNA Mimic > 5 nmol

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.