missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ GAPDH siRNA (human)

Ambion™ Silencer™ GAPDH Positive Control siRNA (human) is ideal for developing and optimizing siRNA experiments.

301.00 € - 722.00 €

Specifications

RNAi Type siRNA
Species Human
Label or Dye Unconjugated
Product Type siRNA
Purity HPLC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10469024
View Documents
Invitrogen™
AM4605
5 nmol
301.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10609845
View Documents
Invitrogen™
AM4633
40 nmol
722.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ambion™ Silencer™ GAPDH Positive Control siRNA (human) is ideal for developing and optimizing siRNA experiments. The control is validated for use in human cell lines. 5 nmol is provided, along with an additional tube of 2 nmol negative control siRNA.

  • Validated siRNA controls for optimizing siRNA experiments
  • Gene-specific control siRNAs provided with scrambled, negative controls
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs.

If you are using other siRNA products, such as Ambion™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.

Specifications

Specifications

siRNA
Unconjugated
HPLC
Human
siRNA
GAPDH
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.