missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ GAPDH siRNA (human, mouse, rat)

Product Code. 10371325
Click to view available options
Quantity:
40 nmol
5 nmol
5 x 40 nmol
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10371325

Brand: Invitrogen™ AM4632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silencer™ GAPDH siRNA (human, mouse, rat)

Invitrogen™ Silencer™ GAPDH Positive Control siRNA (human, mouse, rat) is ideal for developing and optimizing siRNA experiments. The control is validated for use in human, mouse, and rat cell lines.
  • Validated siRNA controls for optimizing siRNA experiments
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs.
If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The siRNAs are provided dried down in single tubes, along with Nuclease-free Water for resuspension, and should be stored at –20°C.
Control Type Positive Control
RNAi Type siRNA
Species Human, Mouse, Rat
For Use With (Application) RNAi
Label or Dye Unconjugated
Product Line Silencer, Ambion
Product Type siRNA
Purity HPLC
Quantity 5 x 40 nmol
Shipping Condition Room Temperature
Target GAPDH
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.