Learn More
Invitrogen™ Silencer™ Human Cell Cycle Regulation siRNA Library
Proteins that regulate cell division are crucial for controlling cell growth and are therefore potentially important drug targets.
Brand:  Invitrogen™ A30137
Product Code. 15691862
  • 22560.00 € / 1 set
Explore more special offers
Description
Description
Proteins that regulate cell division are crucial for controlling cell growth and are therefore potentially important drug targets. The Silencer™ Human Cell Cycle Regulation siRNA Library includes 4,398 Ambion™ Silencer siRNAs targeting 1,466 genes. These siRNA gene targets include cyclin-dependent kinases, or CDKs, which are crucial for cell cycle progression, regulators of cell cycle progression such as CIP/KIP family proteins and members of the INK4 family of cell cycle inhibitors, proteins in the retinoblastoma family, and DNA replication factors such as the cell division cycle proteins (CDCs). The library was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC). Utilizing Ambion Silencer technology built on stringent bioinformatics to enable high target specificity and siRNA potency, the Silencer Human Cell Cycle Regulation siRNA Library is an ideal solution for focused and cost-effective RNAi screening against cell cycle regulation.
  • Comprehensive—Expedite your cell cycle RNAi functional screens with pre-defined Silencer Human Cell Cycle Regulation siRNA Library
  • Reliable—Increase target screening accuracy with three individual siRNAs per gene target
  • Effective—Reduce off target impacts at low concentrations with proven Ambion Silencer siRNA design
  • Convenient—Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats
Why use the Silencer Human Cell Cycle Regulation siRNA Library?
Together with careful bioinformatics selection of gene targets, Silencer siRNAs are optimized with design and high quality synthesis with guaranteed silencing. The Silencer Human Cell Cycle Regulation siRNA Library contains our pre-designed and when available, validated siRNAs generated with maximum potency and specificity in mind so you can focus on data quality. All Silencer libraries are selected against the latest available databases and resources for up-to-date content. Full siRNA sequence information, as well as the exon targeted and each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID, plus results from any validation experiments performed using the included siRNAs is provided. Please contact us for the complete list of targets.
Silencer siRNA libraries allow you to screen efficiently and maximize data output
Ambion Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human Cell Cycle Regulation siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).
Flexibility to meet your workflow needs
Thermo Fisher Scientific offers a comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4398 total siRNAs targeting 1466 genes plated onto 96-well plates (51 plates)
96-well Plate
Human
RNAi
siRNA Library
1 Set
4398 total siRNAs targeting 1466 genes plated onto 51 x 96-well plates
Store at or below -20°C in a non-frost-free freezer.
siRNA
DNA
Silencer™
Standard
Approved for shipment at Room Temperature or on Wet Ice