missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Silencer™ Negative Control No. 1 siRNA
No significant sequence similarity to mouse, rat, or human gene sequences
Brand: Invitrogen™ AM4636
1073.38 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
- Ambion™ Silencer™ Negative Control #1 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences
- The control has also been tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology
- Validated for use in human, mouse, and rat cells
- HPLC purified, duplexed, and ready to use
- Negative control siRNAs are essential for determining transfection efficiency and to control for the effects of siRNA delivery
- In siRNA screening applications, negative control siRNAs are also critical for setting the hit threshold that determines whether an siRNA is considered to have a positive, negative, or neutral effect in a particular assay
- Store at or below -20°C in a non-frost-free freezer

Specifications
The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. | |
siRNA | |
Transfection, RNAi | |
Silencer™, Ambion™ | |
HPLC | |
Room Temperature |
Negative Control | |
Human, Mouse, Rat | |
Unconjugated | |
siRNA | |
5 x 40 nmol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction