Learn More
Invitrogen™ Silencer™ Negative Control No. 4 siRNA
Ambion™ Silencer™ Negative Control #4 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences.
Brand: Invitrogen™ AM4641
Description
Ambion™ Silencer™ Negative Control #4 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences. The control has also been tested in cell-based screens and proved to have no significant effect on cell proliferation, viability, or morphology. 5 nmol is provided.
- Nontargeting with limited sequence similarity to known genes
- Validated for use in human, mouse, and rat cells
- Functionally proven to have minimal effects on cell proliferation and viability
- HPLC purified, duplexed, and ready to use
Negative control siRNAs are essential for determining transfection efficiency and to control for the effects of siRNA delivery. In siRNA screening applications, negative control siRNAs are also critical for setting the 'hit' threshold that determines whether an siRNA is considered to have a positive, negative, or neutral effect in a particular assay.
Accessory Product: Silencer™ siRNA Screening Control Panel (Cat. No. AM4640)
The Silencer™ siRNA Screening Control Panel includes seven negative control siRNAs and is for those researchers performing siRNA screens that employ hundreds or thousands of siRNAs per experiment. Because negative control siRNAs may have unintended effects in a particular assay, a negative control siRNA should be carefully chosen. The Silencer™ siRNA Screening Control Panel simplifies this process. As an added benefit, a positive control siRNA to human, mouse, and rat KIF11 (Eg5) is included in the panel. Knockdown of KIF11, which encodes a kinesin family motor protein, leads to mitotic arrest. Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Ambion™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
Specifications
50μm | |
Silencer™ Negative Control No. 4 siRNA, 5nmol, Liquid, 50μM Concentration, Unconjugated Dye, siRNA RNAi Type, Negative Control Type, Used for RNAi, Transfection, –20°C Storage Temperature, For Research Use Only | |
siRNA | |
RNAi | |
Silencer™, Ambion™ | |
HPLC | |
Dry Ice |
Store at or below -20°C. Do not store in a frost-free freezer. | |
Negative Control | |
Human, Mouse, Rat | |
Unconjugated | |
siRNA | |
5 nmol | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.