Learn More
Thermo Scientific™ Taq Buffers (10X)
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.
22.59 € - 22.84 €
Specifications
Polymerase | Taq DNA Polymerase |
---|---|
Quantity | 4 x 1.25 mL |
Research Category | PCR |
Shipping Condition | Wet or Dry Ice |
Concentration | 10X |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11859622
|
Thermo Scientific™
B55 |
Taq Buffer without Detergent |
22.63 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11819622
|
Thermo Scientific™
B38 |
Taq Buffer with KCl |
22.84 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11879602
|
Thermo Scientific™
B16 |
Taq Buffer with KCl and MgCl2 |
22.62 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11899612
|
Thermo Scientific™
B33 |
Taq Buffer with (NH4)2SO4 |
22.59 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11809622
|
Thermo Scientific™
B34 |
Taq Buffer with (NH4)2SO4 and MgCl2 |
22.65 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA polymerases (both recombinant and native). They are available in different compositions and also without detergents.
10X Taq Buffer without Detergent
- 100 mM Tris-HCl (pH 8.8 at 25°C)
- 500 mM KCl
10X Taq Buffer with KCl and 15 mM MgCl2
- 100 mM Tris-HCl (pH 8.8 at 25°C)
- 500 mM KCl
- 15 mM MgCl2
- Detergent
10X Taq Buffer with (NH4)2SO4
- 750 mM Tris-HCl (pH 8.8 at 25°C)
- 200 mM (NH4)2SO4
- 0.1% (v/v) Tween 20
10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2
- 750 mM Tris-HCl (pH 8.8 at 25°C)
- 200 mM (NH4)2SO4
- 0.1% (v/v) Tween 20
- 20 mM MgCl2
10X Taq Buffer with KCl
- 100 mM Tris-HCl (pH 8.8 at 25°C)
- 500 mM KCl
- Detergent
Specifications
Taq DNA Polymerase | |
PCR | |
10X |
4 x 1.25 mL | |
Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.