Learn More
Molecular Probes™ IANBD Amide (N,N'-Dimethyl-N-(Iodoacetyl)-N'-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)Ethylenediamine)
IANBD Amide (N,N'-Dimethyl-N-(Iodoacetyl)-N'-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)Ethylenediamine)
Brand:  Molecular Probes™ D2004
Product Code. 11520246
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The thiol-reactive IANBD amide can be used to create environment sensitive probes. The absorption and fluorescence emission spectra, quantum yields, and extinction coefficients of NBD conjugates are all markedly dependent on solvent. Additionally, NBD is also a function analog of the dinitrophenyl hapten, and its fluorescence is quenched upon binding to anti-dinitrophenyl (anti-DNP) antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
NBD
IANBD Amide
Thiol
Alkyl Halide, Iodoacetamide
Classic Dyes