missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Cards

Card for use with iBind™ Flex Western Blot Device.

289.00 € - 912.00 €

Specifications

Capacity 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method Chromogenic, Chemiluminescence, Fluorescence
For Use With (Application) Western Blot
For Use With (Equipment) iBind Flex Western Device
Membrane Compatibility Nitrocellulose, PVDF
View More Specs

Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15857696
View Documents
Invitrogen™
SLF2010X4
40 Cards (10 x 4)
912.00 €
Pack of 40
In Stock
to see stock.
Add to basket
 
15367238
View Documents
Invitrogen™
SLF2010
10 Cards
289.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Glass fiber backed card with filer paper stack at base
  • 10 cards per box
Specifications

Specifications

1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Western Blot
Nitrocellulose, PVDF
∼2.5 hr.
Card
Chromogenic, Chemiluminescence, Fluorescence
iBind Flex Western Device
iBind Flex
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots/Card
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ iBind™ Flex Cards

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.