missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Western Device

An automated Western-processing device that performs every step from blocking to washes to antibody incubations via sequential lateral flow

1220.00 € - 3665.00 €

Specifications

Content And Storage Store at room temperature.
Capacity 1 Mini Blot
Detection Method Fluorescence, Chemiluminescence, Chromogenic
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
15120839
View Documents
Invitrogen™
SLF1000
1 device
1220.00 €
Each
In Stock
to see stock.
Add to basket
 
15180839
View Documents
Invitrogen™
SLF10004PK
4 devices
3665.00 €
Pack of 4
In Stock
to see stock.
Add to basket
 
Description

Description

Sequential lateral flow (SLF) allows timely release and flow of solutions and antibodies to the membrane without need of an external power source. Each solution is released from the iBind device wells to an iBind Card via SLF; the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

  • Simple, automated solution
  • No external power source
  • Superior reproducibility over traditional manual methods
  • Better antigen antibody interaction—solutions flow between the iBind Card and membrane for maximum interaction between protein and antibody
  • Minimizes hands-on time
  • Highly reproducible and compatible with all Western protocols
  • Set up time is fast—ready for the detection step within 2.5 hr.
  • Carry out detection right away or wait until the next day without affecting performance
  • Works exclusively with iBind Cards and the iBind Solution Kit (available separately) to process one mini-membrane for immunodetection
  • Provides automated convenience and superior reproducibility over traditional manual methods

Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Specifications

Store at room temperature.
Fluorescence, Chemiluminescence, Chromogenic
Nitrocellulose, PVDF
∼2.5 hr.
Western Device
1 Mini Blot
Western Blot
iBind
1 Mini Blot
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • iBind Western Device

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formamide

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.