Learn More
Ibidi ibiSeal
ibiSeal is a self-adhesive cover film for sealing open wells and reservoirs.
Brand:  Ibidi 10875
Encompass_Preferred
 View more versions of this product
Product Code. 17804794
  • 124.63 € / Pack of 30
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A self-adhesive cover film for sealing open wells and reservoirs in cell culture and microscopy applications
  • Various Dimensions Available
  • sterilized
Specifications
Specifications
4.2 in.
2.8 in.
106.8 mm
Cling Film
71.0 mm
30