Learn More
Ibidi ibiSeal
ibiSeal is a self-adhesive cover film for sealing open wells and reservoirs.
Brand:  Ibidi 10877
Encompass_Preferred
 View more versions of this product
Product Code. 17884784
  • 124.63 € / Pack of 120
Estimated Shipment: 25-09-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A self-adhesive cover film for sealing open wells and reservoirs in cell culture and microscopy applications
  • Various Dimensions Available
  • sterilized
Specifications
Specifications
0.31 in.
0.31 in.
8 mm
Cling Film
8 mm
120