missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ Transfer Stack, nitrocellulose, regular size
GREENER_CHOICE

iBlot Transfer Stacks are used to transfer proteins.

267.00 € - 556.00 €

Specifications

Length (Metric) 13.5 cm
Width (Metric) 8 cm
Format Transfer Stack
Material Nitrocellulose
Pore Size 0.2 μm
View More Specs

Products 2
Product Code Brand Dimensions (LxW) Quantity Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Quantity Price Quantity & Availability  
12023765
GREENER_CHOICE
View Documents
Invitrogen™
IB301031
Regular, 13.5 cm x 8 cm 3 x 10 sets
556.00 €
Pack of 3
In Stock
to see stock.
Add to basket
 
12289309
GREENER_CHOICE
View Documents
Invitrogen™
IB301001
Regular, 13.5 cm x 8 cm 10 sets
267.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

The iBlotTransfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlotTransfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.

Nitrocellulose membrane

  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Bind proteins due to hydrophobic and electrostatic interactions
  • Pore size: 0.2μm
  • Protein binding capacity: 209μg/cm2

 

Transfer stack sizes

  • The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
  • The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)

 

Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Specifications

13.5 cm
Transfer Stack
0.2 μm
iBlot
8 cm
Nitrocellulose
Regular, 13.5 cm x 8 cm
Sustainable disposal
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • iBlot Transfer Stack, nitrocellulose, regular size

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethylene glycol

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.