missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBright™ Prestained Protein Ladder

Product Code. p-7225562
Click to view available options
Quantity:
2 x 250 μL
8 x 250 μL
Unit Size:
Pack of 2
Pack of 4
This item is not returnable. View return policy

Product Code. 15673829

Brand: Invitrogen™ LC5615

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For visualization and protein sizing during Electrophoresis and confirmation of Western blot detection.

Invitrogen iBright Prestained Protein Ladder provide direct visualization during electrophoresis and transfer of Western blots, and chemiluminescent or fluorescent Western blot visualization of two unstained proteins for confirmation of western blot detection.

The iBright Prestained Protein Ladder contains twelve recombinant proteins, ten (11 to 250 kDa) that are blue-stained and fluor-labeled for direct and near-IR fluorescent visualization and protein sizing, and two proteins (30 kDa and 80 kDa) that are unstained and contain IgG binding sites to bind the primary and secondary antibodies used for chemiluminescent and fluorescent detection of the target protein. These two unstained proteins are compatible with chemiluminescent substrates and fluorescent antibodies.

  • Broad size range: twelve proteins spanning 11 to 250 kDa
  • Fluorescent visualization: detect the ten stained proteins using the 670nm red laser or 700nm channel of a fluorescence imager
  • Western blot confirmation: detect the two unstained proteins using the detection system for the target protein
  • Ready to use: supplied in a loading buffer for direct loading on gels
  • Sharp bands: clear, sharp bands for direct visualization on both gel and membrane
  • Reference band: 55 kDa band has greater intensity for easy orientation
  • Quality tested: each lot evaluated by SDS-PAGE and western blotting

Warranty

1 year

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Upon receipt, store at –20°C. Product is shipped on ice.
Detection Method Colorimetric, NIR Fluorescence, and User defined detection system (2 bands)
Number of Markers 12
Product Type Protein Ladder
Ready to Load Yes
Size Range 11 to 250 kDa
Stain Type 1 color: Blue
Gel Compatibility Novex™ Midi Gels, Novex™ Mini Gels, Novex™Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Gels, NuPAGE™ MES Gels, NuPAGE™ MOPS Gels, NuPAGE™ Tris-Acetate Gels, Precise™ Tris-Glycine Gels, SDS-PAGE Gels, Precise™ Tris-HEPES Gels
Molecular Weight (g/mol) 250, 130, 95, 80, 70, 55, 43, 34, 30, 26, 15, 11 kDa
Product Line iBright
Quantity 2 x 250 μL
Shipping Condition Dry Ice
System Type Western Blotting, SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.