Learn More
Invitrogen™ iBright™ System Tray Adapter for E-Gel™ Agarose Gels
Imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.
Brand:  Invitrogen™ A56600
 View more versions of this product
Product Code. 17806521
  • 158.00 € / Each
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Invitrogen iBright Tray Adapters for E-Gel Agarose Gels are imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.
Features of Invitrogen iBright Tray Adapters for E-Gel Agarose Gels include:
• The bottom component centers the E-Gel agarose gels on the iBright transilluminator box facilitating optimized zoom
• The top component covers labels and bar codes on the E-Gel cassette to reduce stray light during image capture and improve sample signal optimization
• Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
• Compatible with 11- to 96-well E-Gel agarose (Cat. No. A56600 for 11-/22-well, Cat. No. A56599 for 48-/96-well)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
iBright
1 Adaptor
E-Gel Tray Adapter, 11-/22-well
Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
Room Temperature