missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ iBu-dG Phosphoramidite

Product Code. 17997192 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17997192

Brand: Thermo Scientific™ 27173405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis.

DNA phosphoramidites with standard base protection ensure a high level of quality and reliability for oligonucleotide synthesis.

iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C44H54N7O8P
Base: dG
Base Protection: iBu
Formula weight: 839.92 g/mol
CAS number: 93183-15-4
HPLC Purity: ≥98.0%
31P NMR Purity: ≥98.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type iBu-dG CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxyguanosine (dG)
Shipping Condition Room Temperature
Purity 98%
Quantity 5 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt