missing translation for 'onlineSavingsMsg'
Learn More

Ibudilast, Tocris Bioscience™

Product Code. 18762644
10 mg
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PDE inhibitor (non-selective)

Phosphodiesterase inhibitor (IC50 values are 53, 35, 48, 12 and 10 μM for PDE Ia, II, III, IV and V respectively). Inhibits platelet aggregation and is an orally-active cerebral vasodilator, bronchodilator and antiallergic agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50847-11-5
Molecular Formula C14H18N2O
Quantity 10 mg
Synonym ibudilast, ketas, ibudilastum, ke tas, ibudilastum latin, ibudilast inn:jan, eyevinal, unii-m0tth61xc5, ketas tn, 3-isobutyryl-2-isopropylpyrazolo 1,5-a pyridine
InChI Key ZJVFLBOZORBYFE-UHFFFAOYSA-N
SMILES CC(C)C1=NN2C=CC=CC2=C1C(=O)C(C)C
IUPAC Name 2-methyl-1-(2-propan-2-ylpyrazolo[1,5-a]pyridin-3-yl)propan-1-one
Molecular Weight (g/mol) 230.311
PubChem CID 3671
Formula Weight 230.31
Percent Purity >99%
Chemical Name or Material Ibudilast
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ibudilast, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.