Learn More
IKA™ ICC Basic Circulator
GREENER_CHOICE
ICC basic circulator with an integrated carrying handle that allows for comfortable transport and safe storage.
Brand:  IKA™ 0004134400
Product Code. 15449783
  • 1499.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Temperature range: RT to 150°C
  • Temperature stability: ± 0.02 K
  • Powerful, speed-controlled pressure/suction pump
  • Holding clamp for attaching the circulator to a bath is included
  • Convenient carrying handle und integrated brackets
  • Large easy to read LED display for pump speed and temperature
  • Push and turn buttons for intuitive menu navigation
  • Adjustable safety circuit
  • Visual and acoustic safety warnings
  • USB / RS 232 interface for PC control and documentation using labworldsoft™, as well as for online firmware updates