Learn More
Sampling Systems™ Ice Cream Sampler Test Tube
Sampling Systems™ Ice Cream Sampler Test Tube offers a place to store the sample taken by an Ice Cream Sampler.
Brand:  Sampling Systems™ 6006A-101
Product Code. 15809381
  • 3.60 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Ice Cream Sampler
  • Clear polystyrene test tube
  • Designed to be easily dismantled for cleaning
Specifications
Specifications
Polystyrene
PS Test Tube
Ice Cream Sampler