Learn More
Scotsman™ Ice Flake Maker Ef156-Install Self Contained
Click to view available options
Capacity (Metric):
60 kg
Description
Ice flakes are extruded at a temperature just below zero degrees and contain 20% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
- Produces granular shape ice flakes, 80% hardness
- Up to 160kg (352 lbs) daily production
- Up to 60kg (132 lbs) ice storage capacity
- Ice scoop holder safely positioned in ice-storage bin
- Resistant stainless steel exterior
- Advanced diagnostics computerised control
- Front panel in & out airflow (air-cooled model only) for built-in installation
- Front access condenser air filter, removable and washable (air-cooled version only)
- Easily accessible, front panel mount, ON-OFF switch
- Easy, front panel access to technical / service area of the machine
- 'Routine Maintenance' visible alarm light on front panel
- Ergonomically designed ice storage access, with disappearing door
- Door-closing movement dampening system
- Vapour-based microbial control system pouch in ice storage area
- Built -in PWD (pumped waste discharge) system for remote water drain (via easy-fit installation) for discharge of waste up to 1.5m above base or up to 20m from unit
- AgION antimicrobial agent embedded in moulded plastics parts
- CFC-free R-404A refrigerant
- Two years manufacturer warranty on defective components
Specifications
Specifications
| Capacity (Metric) | 60 kg |