missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iclaprim, MedChemExpress
MedChemExpress Iclaprim is a new selective bacterial Dihydrofolate inhibitor, which can inhibit the growth of S. aureus (MRSA) with an MIC90 of 0.06 μg/mL.
238.95 € - 6296.40 €
Specifications
Chemical Name or Material | Iclaprim |
---|---|
CAS | 192314-93-5 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C19H22N4O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16453388
|
Medchem Express
HY-101479 |
10 mg |
1194.75 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16439916
|
Medchem Express
HY-101479 |
50 mg |
3905.55 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430769
|
Medchem Express
HY-101479 |
1 mg |
238.95 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16424626
|
Medchem Express
HY-101479 |
100 mg |
6296.40 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16416117
|
Medchem Express
HY-101479 |
5 mg |
716.85 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iclaprim is a new selective bacterial Dihydrofolate inhibitor, which can inhibit the growth of S. aureus (MRSA) with an MIC90 of 0.06 μg/mL.Specifications
Iclaprim | |
Off-White | |
C19H22N4O3 | |
DMSO : 30 mg/mL (84.65 mM; Need ultrasonic and warming) | |
354.4 | |
99.49% | |
Solid |
192314-93-5 | |
Research | |
AR-100 | |
NC1=NC=C(CC2=C3C=CC(C4CC4)OC3=C(OC)C(OC)=C2)C(N)=N1 | |
354.4 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only