missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Tuning Standard solution for 6020 CLP-M, Specpure™, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Alfa Aesar 040482.AE

158.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15436325

  • 158.79 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ICP-MS Tuning Standard solution for 6020 CLP-M
100 mL
UN3264
Liquid
Matrix: 2% HNO3
MFCD00240453
Specpure™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only