Learn More
Indenti-Plug Plastic Foam Stoppers
Keep the contents of your containers secure with these Plastic Foam Stoppers.
Brand: Indenti-Plug 06298-76
This item is not returnable.
View return policy
Description
- Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure.
- Open-cell foam has a uniform density.
- Leaves no fibers like cotton
- Breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use.
- Can also be used as a cleaner for pipettes, tubes, vials and precision tubing.
- Autoclavable.
Specifications
27 to 34 mm | |
44.5 mm | |
Autoclavable | |
Plastic |
Cleaner for pipettes, tubes, vials and precision tubing | |
200 Pk. | |
White | |
Plug Stopper |