Learn More
IDF-11774, MedChemExpress
MedChemExpress IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.
Brand: Medchem Express HY-111387
This item is not returnable.
View return policy
Description
IDF-11774 is a novel hypoxia-inducible factor α (HIFα)-1 inhibitor with an IC50 of 3.65 μM.Specifications
IDF-11774 | |
Off-White | |
C23H32N2O2 | |
DMSO : 60 mg/mL (162.82 mM; Need ultrasonic and warming) | |
368.51 | |
95.37% | |
Solid |
1429054-28-3 | |
Research | |
10 mg | |
CN1CCN(C(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)CC1 | |
368.51 | |
Research | |
Cancer-Kinase/protease |
Research purposes only