Learn More
WTW™ IDS Conductivity Cell TetraCon™ 925/LV with Fork Design
The WTW™ IDS Conductivity Cell 925/LV with Fork Design has proven TetraCon™ technology and is ideal for small samples such as pastes.
Brand:  WTW™ 301719
Product Code. 16232431
  • 836.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • IDS conductivity measurement
  • Proven four-electrode technology
  • Automatic transfer of cell constant and temperature compensation
  • Particularly suited for small samples (ca. 2.5 mL) and paste-like samples
  • Cell constant 0.469 cm-1
  • Fork design
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IDS conductivity cell with four graphite electrodes, epoxy shaft
Small Sample Measurement (2.0.5 mL), Especially Pastes
Plug head for AS/IDS-x cables or wireless modules IDS WLM-S