missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN gamma Rat ELISA Kit

Measure levels of rat interferon gamma (IFNg) in rat serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples using 96-well plates.

Brand:  Invitrogen™ ERIFNG2

 View more versions of this product

Product Code. 10750625

  • 1020.00 € / 1 set
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Assay IFN-γ(rat interferon gamma) levels in rat serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples with our Thermo Scientific Rat IFN gamma ELISA Kits.

These rat IFN-gamma ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interferon gamma (IFNγ) in samples at 50 per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant rat interferon gamma (IFNγ)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 8 to 500pg/mL

  • Sensitivity: < 2pg/mL

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 2.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IL-1α, IL-1β, IL-2, IL-4, IL-6, IL-10, MIP-1α, RANTES, or TNFα, or human or pig IFNγ. When preparations of NIH/NIBSC mouse IFNγ (lot Gg02-901-533) were evaluated in the Rat IFNγ ELISA Kit, 6250pg/mL mouse IFNγ was equivalent to 258pg/mL rat IFNγ (4.1% cross-reactivity)

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

P01581
<2pg/mL
Biotin
ELISA Kit
No significant cross-reaction with rat IL-1a, IL-1ß, IL-2, IL-4, IL-6, IL-10, MIP-1a, RANTES, or TNFa, or human or pig IFNg. When preparations of NIH/NIBSC mouse IFNg (lot Gg02-901-533) were evaluated in the Rat IFNg ELISA Kit, 6250pg/mL mouse IFNg was equivalent to 258pg/mL rat IFN (4.1% cross-reactivity).
Colorimetric Microplate Reader
IFNG2
8.5%
HRP
2 x 96 Tests
Plasma,50μL; Serum,50μL; Supernatant,50μL
Rat
2 hr. 30 min.
8 to 500pg/mL
Rat IFN gamma ELISA Kit
Colorimetric ELISA (TMB substrate) with Plates
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
2 x 96 microplate wells
25712
8.7%
30X Wash Buffer, 2 x 50mL, Adhesive Plate Covers, 12 ea, Anti-Rat IFN gamma 96-well Strip Plate, 2 ea, Biotinylated Antibody Reagent, 2 x 12mL, HRP Dilution Buffer, 2 x 13mL, Lyophilized Recombinant Rat IFN gamma Standard, 4 vials, TMB Substrate Solution, 2 x 12mL, Streptavidin-HRP Concentrate, 2 x 75µL, Stop Solution, 2 x 13mL, Sample Diluent, 2 x 12mL
Absorbance at 450nm minus absorbance at 550nm
RUO
2°C to 8°C
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.