Learn More
Invitrogen™ IgE Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMIGHE
Description
The Mouse Immunoglobulin E (Ms IgE) ELISA quantitates Ms IgE in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IgE. Principle of the method The Mouse IgE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.Specifications
N/A | |
0.14 ng/mL | |
IgE ELISA Kit, 96 Assays, 1 Kit, Mouse Species, Other Proteins Protein Family, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of IgE in Samples | |
96 | |
Colorimetric | |
Other Proteins | |
This ELISA pair antibody detects mouse IgE. Other species not determined. | |
96 Assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
Mouse | |
<10% | |
HRP | |
RUO | |
Plasma, 2 μL; Serum, 2 μL; Supernatant, 100 μL | |
N/A | |
1 hr. 20 min. |
0.137-100 ng/mL | |
0.14ng/mL to 100ng/mL | |
Mouse IgE | |
Biotin | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
TMB | |
Mouse | |
Colorimetric Microplate Reader | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Anti-Mouse IgE precoated 96-well strip plate, lyophilized recombinant mouse IgE standard, 20X wash buffer, biotinylated antibody reagent, streptavidin-HRP reagent, TMB substrate, stop solution, contains 0.2M sulfuric acid, assay diluent (5X concentrated b | |
96 Tests | |
Immunology | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |