missing translation for 'onlineSavingsMsg'
Learn More
IGEPAL∣r CA-630, Thermo Scientific Chemicals Product Code.: 15453719

IGEPAL∣r CA-630, Thermo Scientific Chemicals

Código de producto. 15453719
500 mL
Click to view available options
Quantity:
100 mL
500 mL
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto contiene una sustancia incluida en el anexo XIV del Reglamento REACH (CE) no 1907/2006, todos los clientes deben completar una declaración antes de la entrega.

Product Code. 15453719

Brand: Thermo Scientific Alfa Aesar J61055.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto contiene una sustancia incluida en el anexo XIV del Reglamento REACH (CE) no 1907/2006, todos los clientes deben completar una declaración antes de la entrega.

IGEPAL∣r CA-630, CAS # 9002-93-1, is a nonionic and non-denaturing detergent used mainly for the solubilization, isolation, and purification of membrane protein complexes.

Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins, IGEPAL™ CA-630 is used as a detergent in paper industry, hand cleaners, metal cleaners. It is suitable for solubilization, isolation and purification of membrane protein complexes and in two-dimensional gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • IGEPAL|r CA-630 is a nonionic and non-denaturing detergent similar to Nonidet P-40

Application

  • IGEPAL|r CA-630 is mainly used for the solubilization, isolation, and purification of membrane protein complexes and in two-dimensional gel electrophoresis
  • IGEPAL|r CA-630 can be used in the composition of homogenizing buffers during western blotting technique in mouse muscle homogenates
  • It has been used in X-gal staining to study the lacZ expression
  • IGEPAL|r CA-630 is also a component of the cell lysis buffer in order to prepare RT-qPCR-ready cell lysates
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Octylphenyl-polyethylene glycol
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Especificaciones

Color Colorless
Physical Form Liquid
Density 1.060
Quantity 500 mL
Solubility Information Miscible with water.
Chemical Name or Material IGEPAL™ CA-630

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP25L032 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog NumberJ61055.AE,J61055.AP
Lot NumberQ18K026 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog NumberJ61055.AE,J61055.AP
Lot NumberY25G001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ61055.AE,J61055.AP
Lot NumberM29G014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ61055.AE,J61055.AP
Lot NumberJ17Y018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ61055.AE,J61055.AP
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • IGEPAL CA-630
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contains : IGEPAL CA-630

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
IGEPAL∣r CA-630, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado