missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15699-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is one of six similar proteins that bind insulin-like growth factors I and II (IGF-I and IGF-II). The encoded protein can be secreted into the bloodstream, where it binds IGF-I and IGF-II with high affinity, or it can remain intracellular, interacting with many different ligands. High expression levels of this protein promote the growth of several types of tumors and may be predictive of the chances of recovery of the patient. Several transcript variants, one encoding a secreted isoform and the others encoding nonsecreted isoforms, have been found for this gene.Specifications
IGFBP2 | |
Polyclonal | |
Unconjugated | |
IGFBP2 | |
IGFBP2 | |
IGFBP2 Fusion Protein Ag8327 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18065 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3485 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction